missing translation for 'onlineSavingsMsg'
Learn More

Laurolitsine hydrochloride, MedChemExpress

Product Code. 16499288
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499288 10 mg 10mg
16482589 1 mg 1mg
16431718 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499288 Supplier Medchem Express Supplier No. HYN2352A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Laurolitsine hydrochloride is an alkaloid isolated from Phoebe formosana, and shows weak anti-inflammatory activity.

Laurolitsine hydrochloride is an alkaloid isolated from Phoebe formosana, and shows weak anti-inflammatory activity.

Specifications

Chemical Name or Material Laurolitsine hydrochloride
Color White
Purity Grade Notes Research
Molecular Formula C18H20ClNO4
Quantity 10 mg
Synonym (+)-Norboldine hydrochloride
Solubility Information DMSO : 83 mg/mL (237.27 mM; Need ultrasonic and warming)
SMILES OC1=C(OC)C2=C3C(CCN[C@@]3([H])CC4=CC(O)=C(OC)C=C24)=C1.[H]Cl
Molecular Weight (g/mol) 349.81
Formula Weight 349.81
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.