missing translation for 'onlineSavingsMsg'
Learn More

Lauric Acid, 98-101%, Extra Pure, Acidimetric, SLR, Fisher Chemical

Product Code. 10497640 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10497640 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10497640 Supplier Fisher Chemical Supplier No. L/0560/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 143-07-7
Melting Point 44°C
Color White
Boiling Point 225°C
Packaging Plastic powder jar
Molecular Formula C12H24O2
MDL Number 2736
Quantity 500 g
Synonym lauric acid, n-dodecanoic acid, dodecylic acid, vulvic acid, laurostearic acid, dodecoic acid, duodecylic acid, 1-undecanecarboxylic acid, aliphat no. 4, neo-fat 12
InChI Key POULHZVOKOAJMA-UHFFFAOYSA-N
SMILES CCCCCCCCCCCC(=O)O
IUPAC Name dodecanoic acid
Molecular Weight (g/mol) 200.322
PubChem CID 3893
ChEBI CHEBI:30805
Formula Weight 200.32g/mol
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.