missing translation for 'onlineSavingsMsg'
Learn More

Lauramine Oxide, TRC

Product Code. 30479364
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479364 25 g 25g
30467601 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30479364 Supplier Toronto Research Chemicals Supplier No. TRCL17853025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1643-20-5
Molecular Formula C14 H31 N O
Molecular Weight (g/mol) 229.4
Synonym Dodecylamine, N,N-dimethyl-, N-oxide (6CI,8CI), 101CG, AX, AX (amine oxide), AX 22, AX agent, AX-CS, AX-S, AX-SS, Admox 12, Amiphitol 20N, Ammonyx AO, Ammonyx C10 Amine Oxide, Ammonyx DMCD 40, Ammonyx LO, Ammonyx LO-E, Amogen AOL, Amphitol 20N, Aromox DM 12D, Aromox DM 12D-W, Aromox DM 12DW(C), Aromox DM 12W, Aromox DMA 2DW, Aromox DMCD, Atlas CD 413, Barlox 12i, C 12AX, CFS 560, Cadenac DM 12D-W, Cadenax DM 12DW, Cadenax DM 12DW(C), Cadenax DM 12DW(S), Cadenax DM 12DW-S, ColaLux Lo, Conco XAL, Cyclomox L, DDAO, DDNO, Dimethylaurylamine oxide, Dimethyldodecylamine N-oxide, Dimethyldodecylamine oxide, Dimethyllaurylamine oxide, Dodecydimethylamine-N-oxide, Dodecyldimethylamine oxide, Emal 20N, Emcol L, Emcol LO, Empigen OB, Genaminox K 12, Genaminox LA, Incromine oxide L, LDAO, Lauramine oxide, Laurylamine oxide, Lauryldimethylamine N-oxide, Lauryldimethylamine oxide, Lauryldimethylammonium oxide, MON 59121, Macat AO 12, Mackamine CO-SP, Mackamine LO, N,N-Dimethyl-1-dodecanamine N-oxide, N,N-Dimethyl-1-dodecanamine oxide, N,N-Dimethyl-N-dodecylamine N-oxide, N,N-Dimethyl-n-dodecylamine oxide, N,N-Dimethyldodecylamine N-oxide, N,N-Dimethyldodecylamine oxide, N-Dodecyl-N,N-dimethylamine oxide, N-Lauryl-N,N-dimethylamine oxide, N-Lauryldimethylamine N-oxide, Ninox DMCD 40, Ninox L, Nissan Unisafe A-LM, OA 12, OB 2, OB 2 (surfactant), Oxamin LO, Oxidet DM 20, Rewominox L 408, Rhodamox L, Rhodamox LO, Schercamox DML, Softamine L, TAO 14, Tomah AO 728, Unisafe A-LM, Unisafe A-LMR, Unisorb A-LM, Vicamox 401, Wondamin OX 300, Wondamine OX 300, n-Dodecyl-N,N-dimethylamine-N-oxide, n-Dodecyldimethylamine oxide, 1-Dodecanamine, N,N-dimethyl-, N-oxide
IUPAC Name N,N-dimethyldodecan-1-amine oxide
SMILES CCCCCCCCCCCC[N+](C)(C)[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C14H31NO/c1-4-5-6-7-8-9-10-11-12-13-14-15(2,3)16/h4-14H2,1-3H3
Formula Weight 229.2406
Percent Purity >95%
Chemical Name or Material Lauramine Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.