missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LATS2, GST Tag Recombinant Protein

Product Code. 15409003
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15409003

Brand: Invitrogen™ PV6365

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human LATS2 (480-1088) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for LATS2 is NP_055387.2.

LATS2 is a serine/threonine protein kinase belonging to the LATS tumor suppressor family. LATS2 interacts with a negative regulator of p53 and function in a positive feedback loop with p53 that responds to cytoskeleton damage and this interaction provokes centrosome/mitotic apparatus dysfunction. LATS2 plays an essential role in the maintenance of mitotic fidelity and genomic integrity. LATS2 plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1, which in turn phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. Phosphorylation of YAP1 by LATS2 inhibits its translocation into the nucleus to regulate cellular genes important for cell proliferation, cell death, and cell migration. It negatively regulates G1/S transition by down-regulating cyclin E/CDK2 kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NRM7
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 26524
Molecular Weight (g/mol) 95.6 kDa
Name Human LATS2, GST Tag
Quantity 100 μg
Gene Alias 4932411G09Rik; AV277261; AW228608; h-warts; Kinase phosphorylated during mitosis protein; KPM; large tumor suppressor 2; large tumor suppressor homolog 2; large tumor suppressor kinase 2; large tumor suppressor, homolog 2; LATS (large tumor suppressor, Drosophila) homolog 2; LATS, large tumor suppressor, homolog 2; LATS2; serine/threonine kinase KPM; serine/threonine-protein kinase kpm; Serine/threonine-protein kinase LATS2; si:ch211-173o4.2; Warts-like kinase
Common Name LATS2
Protein Length 480-1088
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.