missing translation for 'onlineSavingsMsg'
Learn More

LATS1 Antibody, Novus Biologicals™

Product Code. 18386773 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386773 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386773 Supplier Novus Biologicals Supplier No. NBP305224

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LATS1 Polyclonal specifically detects LATS1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LATS1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EC 2.7.11, h-warts, Large tumor suppressor homolog 1, LATS (large tumor suppressor, Drosophila) homolog 1, LATS, large tumor suppressor, homolog 1 (Drosophila), serine/threonine-protein kinase LATS1, WARTS protein kinase, WARTSEC 2.7.11.1, wts
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human LATS1 (NP_004681.1). EVNPQMLQDLQAAGFDEDMVIQALQKTNNRSIEAAIEFISKMSYQDPRREQMAAAAARPINASMKPGNVQQSVNRKQSWKGSKESLVPQRHGPPLGESVAY
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Mitotic Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9113
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.