missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Latrunculin A

Product Code. 18005152
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18005152 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18005152 Supplier R&D Systems™ Supplier No. 3973/100U
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of actin assembly and polymerization

Latrunculin A is a reversible inhibitor of actin assembly; blocks actin adenine nucleotide exchange. Complexes with actin in vitro and interacts with actin monomers only, unlike cytochalasins. Prevents actin repolymerization into filaments and disrupts the actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 μg
Product Line Tocris
Product Type Latrunculin A
Purity 0.95
Molecular Weight (g/mol) 421.55
Inhibitors Latrunculin A

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.