missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Latrophilin 2/LPHN2 Antibody (1030202) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB105881JF669
This item is not returnable.
View return policy
Description
Latrophilin 2/LPHN2 Monoclonal antibody specifically detects Latrophilin 2/LPHN2 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| Latrophilin 2/LPHN2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Calcium-independent alpha-latrotoxin receptor 2, CIRL2, CIRL-2, CL2, KIAA0786, latrophilin 2, Latrophilin homolog 1, latrophilin homolog 2 (cow), latrophilin-2, LEC1LPHH1latrophilin 1, lectomedin-1 | |
| Human Latrophilin 2/LPHN2 synthetic peptide, Accession # O95490 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, CyTOF | |
| 1030202 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, GPCR | |
| 23266 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction