missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Latexin Antibody (02), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06086H
This item is not returnable.
View return policy
Description
Latexin Monoclonal antibody specifically detects Latexin in Human samples. It is validated for Flow Cytometry
Specifications
| Latexin | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 02 | |
| Flow Cytometry | |
| ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, E.coli-derived, recombinant Human Latexin (Accession#: NP_064554.3; Glu 2-Glu 222). | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 56925 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction