missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Latexin Monoclonal specifically detects Latexin in Mouse samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Latexin |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 9 |
| Conjugate | Alexa Fluor 488 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Latexin (NP_058033.2; Glu 2-Glu 222). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?