missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: Staphylococcus aureus
Shop All Novacyt Group Products
Click to view available options
Quantity:
100 Tests
500 Tests
Unit Size:
100 tests
50 tests
500 tests
Description
- Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
- Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
- Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | S. aureus |
| Type | Latex Agglutination Test |
| Quantity | 100 Tests |
| Test Time | 2 min. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction