missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LATEX 400nm Red Mini Antibody Conjugation Kit

Product Code. 18024516 Shop All Bio Techne Products
Click to view available options
Quantity:
16 μg
40 μg
Unit Size:
16µg
Pack of 10
This item is not returnable. View return policy

Product Code. 18024516

Brand: Novus Biologicals™ 10020100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The LATEX™ Conjugation Kit allows antibodies or proteins to be conjugated to high quality 400 nm red latex beads quickly and easily.

The LATEX™ Conjugation Kit allows antibodies or proteins to be conjugated to our high quality 400nm red latex beads quickly and easily. The latex beads are designed for ease of use and have much improved handling compared with traditional latex. This has been achieved by specially treating the particles. The conjugation is covalent, and requires less antibody than traditional passive and covalent latex conjugates.

Kit Contents:

  • 1 vial of Reaction buffer B
  • 1 vial of Reaction buffer A
  • 1 vial of Resuspension buffer
  • 1 vial of 10x Quencher buffer
  • 4 or 10 Mini vials of 400nm Red LATEX
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 40 μg
Target LATEX™ 400nm Red Mini
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.