missing translation for 'onlineSavingsMsg'
Learn More

Latent TGF-beta bp1 Antibody (MM0447-5L38), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18429747 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429747 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429747 Supplier Novus Biologicals Supplier No. NBP211783MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Latent TGF-beta bp1 Monoclonal antibody specifically detects Latent TGF-beta bp1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Latent TGF-beta bp1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Gene Alias latent transforming growth factor beta binding protein 1, latent-transforming growth factor beta-binding protein 1, LTBP-1, MGC163161, TGF-beta1-BP-1, Transforming growth factor beta-1-binding protein 1
Host Species Mouse
Quantity 0.1 mL
Research Discipline Ovarian Carcinoma Cell Markers
Gene ID (Entrez) 4052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.