Learn More
Invitrogen™ LAT Recombinant Rabbit Monoclonal Antibody (24GB4940)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557444
 View more versions of this product
Product Code. 30276571
  • £488.00 / 100µL
Description
LAT Recombinant Monoclonal Antibody for Western Blot, Flow
LAT (Linker for Activation of T cells) is a 36-38 kDa double-palmitoylated transmembrane adaptor protein expressed by T cells, pre-B cells, NK cells, mast cells and platelets. After immunoreceptor triggering, LAT becomes multiply tyrosine-phosphorylated by Syk-, Src-, or Tec-family kinases, providing docking sites for downstream signaling molecules. LAT is essential for TCR-dependent T cell- and FcepsilonRI-dependent mast cell activation, as well as for maturation of early thymocytes. It is also involved in NK cell signaling and platelet activation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LAT
Recombinant Monoclonal
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O43561
LAT
A synthesized peptide derived from human LAT (aa 217-234).
100 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4940
Unconjugated
LAT
36 kDa phospho-tyrosine adapter protein; 36 kDa phospho-tyrosine adaptor protein; Lat; LAT1; linker for activation of T cells; linker for activation of T cells, transmembrane adaptor; linker for activation of T-cells; linker for activation of T-cells family member 1; linker protein; p36-38; pp36
Rabbit
Affinity chromatography
RUO
27040
-20°C