missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAT Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £348.00
Specifications
| Antigen | LAT |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18370792
|
Novus Biologicals
NBP3-16940-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18385576
|
Novus Biologicals
NBP3-16940-100UL |
100 μg |
£348.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAT Polyclonal antibody specifically detects LAT in Human, Mouse samples. It is validated for Western BlotSpecifications
| LAT | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| A synthetic phosphorylated peptide around Y191 of human LAT (NP_001014987.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 27040 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title