missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582419
This item is not returnable.
View return policy
Description
Immunogen sequence: HCHRLPGSYD STSSDSLYPR GIQFKRPHTV APWPPAYPPV TSYPPLSQPD LLPIPRSPQP LGGSHRTPSS RRDSDG Highest antigen sequence indentity to the following orthologs: Mouse - 67%, Rat - 67%.
LAT (Linker for Activation of T cells) is a 36-38 kDa double-palmitoylated transmembrane adaptor protein expressed by T cells, pre-B cells, NK cells, mast cells and platelets. After immunoreceptor triggering, LAT becomes multiply tyrosine-phosphorylated by Syk-, Src-, or Tec-family kinases, providing docking sites for downstream signaling molecules. LAT is essential for TCR-dependent T cell- and FcepsilonRI-dependent mast cell activation, as well as for maturation of early thymocytes. It is also involved in NK cell signaling and platelet activation.
Specifications
| LAT | |
| Polyclonal | |
| Unconjugated | |
| LAT | |
| 36 kDa phospho-tyrosine adapter protein; 36 kDa phospho-tyrosine adaptor protein; Lat; LAT1; linker for activation of T cells; linker for activation of T cells, transmembrane adaptor; linker for activation of T-cells; linker for activation of T-cells family member 1; linker protein; p36-38; pp36 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27040 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43561 | |
| LAT | |
| Recombinant protein corresponding to Human LAT. Recombinant protein control fragment (Product #RP-90454). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction