missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAT Polyclonal Antibody, Invitrogen™
Description
The synthetic peptide sequence is 242-262aa, QEAEEVEEEGAPDYENLQELN Add 0.2 mL of distilled water, will yield a concentration of 500 μg/mL.
LAT (Linker for Activation of T cells) is a 36-38 kDa double-palmitoylated transmembrane adaptor protein expressed by T cells, pre-B cells, NK cells, mast cells and platelets. After immunoreceptor triggering, LAT becomes multiply tyrosine-phosphorylated by Syk-, Src-, or Tec-family kinases, providing docking sites for downstream signaling molecules. LAT is essential for TCR-dependent T cell- and FcepsilonRI-dependent mast cell activation, as well as for maturation of early thymocytes. It is also involved in NK cell signaling and platelet activation.
Specifications
Specifications
| Antigen | LAT |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 5MG BSA, 5MG BSA and 0.05MG sodium azide, 0.05MG sodium azide, 0.05MG thimerosal, 0.05MG thimerosal |
| Gene | LAT |
| Gene Accession No. | O43561 |
| Gene Alias | Linker for activation of T-cells family member 1, 36 kDa phospho-tyrosine adapter protein, pp36, p36-38, LAT |
| Gene Symbols | LAT |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?