missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27104SS
This item is not returnable.
View return policy
Description
LAT Polyclonal specifically detects LAT in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LAT | |
| Polyclonal | |
| Unconjugated | |
| PBS, 0.05% BSA with 0.05% Sodium Azide | |
| LAT | |
| A portion of amino acid 70-120 of human LAT was used as the immunogen. 86 cQPLGGSHRTPSSRRDS 101 | |
| 0.025 mg | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 27040 | |
| Human, Mouse, Primate | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/mL, Immunohistochemistry-Paraffin | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for the LAT antibody is 100% homologous in human, rhesus monkey and chimpanzee, 88% homologous in new world monkey, 82% homologous in rat, rabbit, dog and pig, and 75% homologous in mouse and cow. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction