missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lassa virus RING finger protein Z Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112046
This item is not returnable.
View return policy
Description
Lassa virus RING finger protein Z Polyclonal Antibody for Western Blot
Plays a crucial role in virion assembly and budding. Expressed late in the virus life cycle, it acts as an inhibitor of viral transcription and RNA synthesis by interacting with the viral polymerase L. Presumably recruits the NP encapsidated genome to cellular membranes at budding sites via direct interaction with NP. Plays critical roles in the final steps of viral release by interacting with host TSG101, a member of the vacuolar protein-sorting pathway and using other cellular host proteins involved in vesicle formation pathway. The budding of the virus progeny occurs after association of protein Z with the viral glycoprotein complex SSP-GP1-GP2 at the cell periphery, step that requires myristoylation of protein Z. Also selectively represses protein production by associating with host eIF4E.
Specifications
| Lassa virus RING finger protein Z | |
| Polyclonal | |
| Unconjugated | |
| Lassa virus | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Lassa virus RING finger protein Z (Lassa virus (strain mouse/Sierra Leone/Josiah/1976)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.11 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction