missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lassa virus RING finger protein Z Antibody, Novus Biologicals™
Description
Lassa virus RING finger protein Z Polyclonal antibody specifically detects Lassa virus RING finger protein Z in Virus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Lassa virus RING finger protein Z |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 20% Glycerol (pH7) |
| Gene Alias | LASV Z, Protein Z, Zinc-binding protein |
| Host Species | Rabbit |
| Immunogen | Full length Lassa virus RING finger protein Z recombinant protein. |
| Purification Method | Protein A purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?