missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lassa Virus Nucleoprotein Recombinant Rabbit Monoclonal Antibody (HL1390)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547025
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Lassa fever virus, a member of the family Arenaviridae, is a highly endemic category A pathogen that causes 300,000-500,000 infections per year in Western Africa. The arenaviral nucleoprotein NP has been implicated in suppression of the host innate immune system.
Specifications
| Lassa Virus Nucleoprotein | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1390 | |
| Unconjugated | |
| Lassa mammarenavirus NP; Lassa mammarenavirus Nucleoprotein; Lassa virus NP; Lassa virus Nucleoprotein; LASV NP; LASV Nucleoprotein | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Lassa virus Nucleoprotein. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction