missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LASS6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30906-100ug
This item is not returnable.
View return policy
Description
LASS6 Polyclonal antibody specifically detects LASS6 in Human samples. It is validated for Western Blot
Specifications
| LASS6 | |
| Polyclonal | |
| Western Blot : 1:400 | |
| CerS6, LAG1 homolog, ceramide synthase 6, LAG1 longevity assurance homolog 6, LAG1 longevity assurance homolog 6 (S. cerevisiae), longevity assurance homolog 6, MGC129949, MGC129950 | |
| Synthetic peptide dervied from human LASS6 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Tris buffered saline, 10mM MgCl2, 10mM ammonium sulfate, and 0.2% lactose | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 253782 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction