missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lass5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA520647

Product Code. 12878792

  • £439.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A suggested positive control is SK-N-SH cell lysate. PA5-20647 can be used with blocking peptide PEP-0767.

The LASS (longevity assurance homolog) family members represent a subgroup of the homeobox gene family and are highly conserved from yeasts to mammals. Six members of this family of proteins have been characterized (LASS1-6) and all are involved in ceramide synthesis during cell growth regulation and cancer differentiation. LASS5, also called Trh4, is a 392 amino acid endoplasmic reticulum, multi-pass membrane protein. Functioning as a dihydro-ceramide synthase, LASS5 is involved in the production of sphingolipids containing mainly one fatty acid donor (N-linked palmitoyl-ceramide) in a fumonisin B1-independent manner. It uses palmitoyl-CoA as an acyl donor and is involved in the synthesis of C14, C16 and C18-ceramide. LASS5 is the most abundantly expressed and predominant ceramide synthase isoform in lung epithelia. Recent studies show that LASS5 partially correct growth and apoptosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lass5
Polyclonal
Unconjugated
CERS5
2310081H14Rik; AU045339; AW544927; Ceramide synthase 5; CerS5; LAG1 homolog, ceramide synthase 5; LAG1 longevity assurance homolog 5; LASS5; longevity assurance homolog 5; Sphingosine N-acyltransferase CERS5; TRAM homolog 4; translocating chain-associating membrane protein homolog 4; Trh4
Rabbit
Antigen affinity chromatography
RUO
366984, 71949, 91012
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8N5B7, Q9D6K9
CERS5
A 14 amino acid peptide near the amino terminus of the human LASS5.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.