missing translation for 'onlineSavingsMsg'
Learn More

Lass5 Antibody, HRP, Novus Biologicals™

Product Code. 18407660 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407660 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407660 Supplier Novus Biologicals Supplier No. NBP177328H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Lass5 Polyclonal antibody specifically detects Lass5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lass5
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias CerS5, FLJ25304, LAG1 homolog, ceramide synthase 5, LAG1 longevity assurance homolog 5, LAG1 longevity assurance homolog 5 (S. cerevisiae), MGC45411, Trh4
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human LASS5. The immunogen is located within the last 50 amino acids of LASS5. Amino Acid Squence: SKDDRSDVESSSEEEDV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91012
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.