missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lass4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA112928
This item is not returnable.
View return policy
Description
A suggested positive control for this product is lung or embryo lysate.
The LASS (longevity assurance homolog) family members are highly conserved from yeasts to mammals. Six members of this family of proteins have been characterized (LASS1, LASS2, LASS3, LASS4, LASS5 and LASS6) and they are all involved in sphingolipid synthesis. LASS4 is a 394 amino acid endoplasmic reticulum, multi-pass membrane protein. LASS4 increases the levels of long ceramides such as C22:0- and C24:0-ceramides. In cells deficient for CLN9, as observed in neuronal ceroid lipofuscinosis (NCL) or Batten disease, LASS4 can increase ceramide levels and partially correct growth and apoptosis.
Specifications
| Lass4 | |
| Polyclonal | |
| Unconjugated | |
| CERS4 | |
| 2900019C14Rik; Ceramide synthase 4; CerS4; FLJ12089; LAG1 homolog, ceramide synthase 4; LAG1 longevity assurance homolog 4; Lass4; longevity assurance homolog 4; Sphingosine N-acyltransferase CERS4; TRAM homolog 1; translocating chain-associating membrane protein homolog 1; Trh1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79603 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.08% sodium azide | |
| Q9HA82 | |
| CERS4 | |
| Synthetic peptide corresponding to human Lass4 protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction