missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lass4 Polyclonal Antibody

Lass4 Polyclonal Antibody
Brand: Invitrogen PA599248
This item is not returnable.
View return policy
Description
Lass4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| Lass4 | |
| Polyclonal | |
| Unconjugated | |
| CERS4 | |
| 2900019C14Rik; Ceramide synthase 4; CerS4; FLJ12089; LAG1 homolog, ceramide synthase 4; LAG1 longevity assurance homolog 4; Lass4; longevity assurance homolog 4; Sphingosine N-acyltransferase CERS4; TRAM homolog 1; translocating chain-associating membrane protein homolog 1; Trh1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79603 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HA82 | |
| CERS4 | |
| A synthesized peptide derived from human CERS4(Accession Q9HA82), corresponding to amino acid residues L49-Q99. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction