missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lass2 Recombinant Rabbit Monoclonal Antibody (HL1717)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547154
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
LASS2 is a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. This gene encodes a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. Alternatively spliced transcript variants encoding the same protein have been described.
Specifications
| Lass2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q96G23 | |
| CERS2 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human LASS2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL1717 | |
| Unconjugated | |
| CERS2 | |
| 0610013I17Rik; AI225939; ceramide synthase 2; CERS2; FLJ10243; L3; LAG1 homolog, ceramide synthase 2; LAG1 longevity assurance 2; LAG1 longevity assurance homolog 2; LAG1 longevity assurance homolog 2; ceramide synthase 2; LASS2; longevity assurance (LAG1, S. cerevisiae) homolog 2; longevity assurance homolog 2; MGC987; SP260; Sphingosine N-acyltransferase CERS2; TMSG1; TRAM homolog 3; translocating chain-associating membrane protein homolog 3; Trh3; Tumor metastasis-suppressor gene 1 protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 29956 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction