missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LASS1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-28480
This item is not returnable.
View return policy
Description
LASS1 Polyclonal specifically detects LASS1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| LASS1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CerS1, LAG1 homolog, ceramide synthase 1, LAG1 longevity assurance homolog 1, LAG1MGC90349, longevity assurance (LAG1, S. cerevisiae) homolog 1, Longevity assurance gene 1 protein homolog 1, Protein UOG-1, UOG1LAG1 longevity assurance homolog 1 (S. cerevisiae), upstream of GDF1 | |
| Goat | |
| 40 kDa | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_067090.1and NP_937850.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| P27544 | |
| CERS1 | |
| Peptide with sequence C-KDLREYDTAEAQ corresponding to internal region according to NP_937850.1. | |
| Affinity Purified | |
| RUO | |
| 10715 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction