missing translation for 'onlineSavingsMsg'
Learn More

Lasalocid, MedChemExpress

Product Code. 16431098
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16431098

Brand: Medchem Express HYB1071

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lasalocid (Lasalocid-A; Ionophore X-537A; Antibiotic X-537A) is an antibacterial agent and a coccidiostat, used in the feed additives

Lasalocid (Lasalocid-A

Specifications

Chemical Name or Material Lasalocid
Physical Form Solid
CAS 25999-31-9
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C34H54O8
For Use With (Application) COVID-19-immunoregulation
Synonym Lasalocid-A Ionophore X-537A Antibiotic X-537A
Solubility Information DMSO : 100 mg/mL (169.26 mM; Need ultrasonic)
SMILES O=C(O)C1=C(CC[C@@H](C)[C@H](O)[C@H](C)C([C@@H]([C@H]2O[C@@]([C@H]3CC[C@](O)(CC)[C@H](C)O3)(CC)C[C@@H]2C)CC)=O)C=CC(C)=C1O
Molecular Weight (g/mol) 590.79
Formula Weight 590.79
Percent Purity 96.85%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.