missing translation for 'onlineSavingsMsg'
Learn More

LAS2 Antibody (OTI2B2), DyLight 594, Novus Biologicals™

Product Code. 18429365 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429365 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429365 Supplier Novus Biologicals Supplier No. NBP272452DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LAS2 Monoclonal antibody specifically detects LAS2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAS2
Applications Western Blot
Classification Monoclonal
Clone OTI2B2
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias C18orf54, chromosome 18 open reading frame 54, hypothetical protein LOC162681, LAS2, MGC33382
Host Species Mouse
Immunogen Full length recombinant protein of human LAS2 (NP_775800) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162681
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.