missing translation for 'onlineSavingsMsg'
Learn More

LAS-34823, TRC

Product Code. 30476300
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
30476300 10 mg 10mg
30466056 100 mg 100mg
30464577 50 mg 50mg
3 options
This item is not returnable. View return policy

Product Code. 30476300

Brand: Toronto Research Chemicals TRCL39503010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1708930-15-7
Formula Weight 341.099
InChI Formula InChI=1S/C16H24NO2.BrH/c18-16-13-17(10-7-14(16)8-11-17)9-4-12-19-15-5-2-1-3-6-15;/h1-3,5-6,14,16,18H,4,7-13H2;1H/q+1;/p-1/t14?,16-,17?;/m0./s1
IUPAC Name (3R)-1-(3-phenoxypropyl)-1-azoniabicyclo[2.2.2]octan-3-ol;bromide
Molecular Formula C16H24BrNO2
Molecular Weight (g/mol) 342.27
SMILES [Br-].O[C@H]1C[N+]2(CCCOc3ccccc3)CCC1CC2
Synonym LAS-34823, (R)-3-Hydroxy-1-(3-phenoxypropyl)-1-azoniabicyclo[2,2.2]octane Bromide, (R)-3-Hydroxy-1-(3-phenoxypropyl)quinuclidium Bromide
Chemical Name or Material Aclidinium Bromide Impurity 2

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.