Learn More
LARS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-32208
Product Code. 18022744
  • £450.00 / 100µL
Description
LARS Polyclonal specifically detects LARS in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LARS
Polyclonal
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
cytoplasmic leucyl-tRNA synthetase, FLJ21788, hr025Cl, HSPC192, KIAA1352, LARS1, leucine translase, leucine tRNA ligase 1, cytoplasmic, leucine-tRNA ligase, leucine--tRNA ligase, leucyl-tRNA synthetase, leucyl-tRNA synthetase, cytoplasmic, LEURS, LEUS, LRS, PIG44
Rabbit
Affinity Purified
RUO
51520
Store at 4C. Do not freeze.
Western Blot, Immunoprecipitation
1.0 mg/mL
Western Blot 1:1000 - 1:5000, Immunoprecipitation
NP_064502.9
LARS
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Leucyl-tRNA Synthetase using the numbering given in entry NP_064502.9 (GeneID 51520).
100 μL
Primary
Human, Mouse