missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LARS Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32208FR
This item is not returnable.
View return policy
Description
LARS Polyclonal antibody specifically detects LARS in Human, Mouse samples. It is validated for Western Blot
Specifications
| LARS | |
| Polyclonal | |
| Western Blot | |
| cytoplasmic leucyl-tRNA synthetase, FLJ21788, hr025Cl, HSPC192, KIAA1352, LARS1, leucine translase, leucine tRNA ligase 1, cytoplasmic, leucine-tRNA ligase, leucine--tRNA ligase, leucyl-tRNA synthetase, leucyl-tRNA synthetase, cytoplasmic, LEURS, LEUS, LRS, PIG44 | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Leucyl-tRNA Synthetase using the numbering given in entry NP_064502.9 (GeneID 51520). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51520 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction