missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LARP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17067-1-AP-20UL
This item is not returnable.
View return policy
Description
LARP7 gene ontology annotations related to this gene include RNA processing.Specifications
| LARP7 | |
| Polyclonal | |
| Unconjugated | |
| LARP7 | |
| HDCMA18P, La related protein 7, LARP7, PIP7S | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 28036, 51574, 686883 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q05CL8, Q4G0J3, Q5XI01 | |
| LARP7 | |
| LARP7 Fusion Protein Ag10891 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction