missing translation for 'onlineSavingsMsg'
Learn More

LARP6 Antibody, Novus Biologicals™

Product Code. 18351449 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351449 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351449 Supplier Novus Biologicals Supplier No. H00055323B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

LARP6 Polyclonal antibody specifically detects LARP6 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen LARP6
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:2000, Immunoprecipitation
Formulation PBS (pH 7.4)
Gene Accession No. NP_060827
Gene Alias Acheron, Achn, death-associated LA motif protein, FLJ11196, La ribonucleoprotein domain family member 6, La ribonucleoprotein domain family, member 6, la-related protein 6
Host Species Mouse
Immunogen LARP6 (NP_060827, 1 a.a. - 491 a.a.) full-length human protein. MAQSGGEARPGPKTAVQIRVAIQEAEDVDELEDEEEGAETRGAGDPARYLSPGWGSASEEEPSRGHSGTTASGGENEREDLEQEWKPPDEELIKKLVDQIEFYFSDENLEKDAFLLKHVRRNKLGYVSVKLLTSFKKVKHLTRDWRTTAHALKYSVVLELNEDHRKVRRTTPVPLFPNENLPSKMLLVYDLYLSPKLWALATPQKNGRVQEKVMEHLLKLFGTFGVISSVRILKPGRELPPDIRRISSRYSQVGTQECAIVEFEEVEAAIKAHEFMITESQGKENMKAVLIGMKPPKKKPAKDKNHDEEPTASIHLNKSLNKRVEELQYMGDESSANSSSDPESNPTSPMAGRRHAATNKLSPSGHQNLFLSPNASPCTSPWSSPLAQRKGVSRKSPLAEEGRLNCSTSPEIFRKCMDYSSDSSVTPSGSPWVRRRRQAEMGTQEKSPGTSPLLSRKMQTADGLPVGVLRLPRGPDNTRGFHGHERSRACV
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55323
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.