missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LARP2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA589091

Product Code. 16527121

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: FDYSYGYQEH GERTDQPFQT ELNTSMMYYY DDGTGVQVYP VEEALLKEYI KRQIEYYFSV ENLERDFFLR GKMDEQGFLP ISLIAGFQRV QALTTNLNLI LEALKDSTEV EIVDEKMRKK IEPEKWPIPG PPPRSVPPTD FSQLIDCPEF V; Positive Samples: A-549, HT-29, HeLa, U-87MG, Mouse brain.

LARP2 encodes a protein containing domains found in the La related protein of Drosophila melanogaster. La motif-containing proteins are thought to be RNA-binding proteins, where the La motif and adjacent amino acids fold into an RNA recognition motif. The La motif is also found in proteins unrelated to the La protein. Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LARP2
Polyclonal
Unconjugated
LARP1B
1700108L22Rik; 4933421B21Rik; AA690246; DKFZp686L13217; fc76b04; la related protein; La ribonucleoprotein 1B; La ribonucleoprotein domain family member 1B; La ribonucleoprotein domain family member 2; La ribonucleoprotein domain family, member 1B; La ribonucleoprotein domain family, member 2; La-like protein 1B-like protein; la-related protein 1; la-related protein 1B; la-related protein 1B; La-related protein 2; LARP; LARP1B; LARP2; MGC117277; MGC75174; RGD1307509; wu:fc76b04
Rabbit
Affinity chromatography
RUO
214048, 310348, 55132
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
0.66 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q659C4
LARP1B
Recombinant fusion protein containing a sequence corresponding to amino acids 170-320 of human LARP1B (NP_0605482).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.