missing translation for 'onlineSavingsMsg'
Learn More

LARP1 Rabbit anti-Drosophila, Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7205962 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16822064 20 μL 20µL
16812064 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16822064 Supplier Proteintech Supplier No. 137081AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNA-binding protein that promotes translation of specific classes of mRNAs downstream of the mTORC1 complex. Associates with the mRNA 5′cap in an MTOR-dependent manner and associates with mRNAs containing a 5′ terminal oligopyrimidine (5′TOP) motif, which is present in mRNAs encoding for ribosomal proteins and several components of the translation machinery. Associates with actively translating ribosomes via interaction with PABPC1/PABP and stimulates translation of mRNAs containing a 5′TOP, thereby regulating cell growth and proliferation. Positively regulates the replication of dengue virus (DENV).

Specifications

Antigen LARP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LARP1
Gene Accession No. Q6PKG0, Q6ZQ58
Gene Alias KIAA0731, La related protein 1, LARP, LARP1
Gene Symbols larp, Larp1
Host Species Rabbit
Immunogen LARP1 Fusion Protein Ag4645
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23367, 303158, 53567, 73158
Target Species Human, Drosophila, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.