missing translation for 'onlineSavingsMsg'
Learn More

Larotrectinib, MedChemExpress

Product Code. 16485408
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485408 10 mg 10mg
16451597 50 mg 50mg
16438127 5 mg 5mg
16426016 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485408 Supplier Medchem Express Supplier No. HY12866

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Larotrectinib (LOXO-101) is an ATP-competitive oral, selective inhibitor of the tropomyosin-related kinase (TRK) family receptors, with low nanomolar 50% inhibitory concentrations against all three isoforms (TRKA, B, and C).

Larotrectinib (LOXO-101) is an ATP-competitive oral, selective inhibitor of the tropomyosin-related kinase (TRK) family receptors, with low nanomolar 50% inhibitory concentrations against all three isoforms (TRKA, B, and C).

Specifications

Chemical Name or Material Larotrectinib
CAS 1223403-58-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H22F2N6O2
Quantity 10 mg
Synonym LOXO-101 ARRY-470
Solubility Information DMSO : ≥ 4.6 mg/mL (10.74 mM)
SMILES O=C(N1C[C@@H](O)CC1)NC2=C3N=C(N4[C@@H](C5=CC(F)=CC=C5F)CCC4)C=CN3N=C2
Molecular Weight (g/mol) 428.44
Formula Weight 428.44
Percent Purity 98.92%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.