missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan HDPE, Large Sealable Barrel
Click to view available options
Plastic Barrel
Description
- Wide opening for ease of access to contents
- Opens and closes quickly without a suction effect
- Hermetic and waterproof seal
- 3-year warranty
- Screw lid closure
- UN-X Group 1 approved for the international transport of hazardous products over land, maritime or air
- Incinerable and 100% recyclable
Specifications
Capacity (English) | 1.5 gal. |
Product Type | Barrel |
Certifications/Compliance | UN X Group 1 |
Description | Temperature range from −40° to +80°C |
Diameter (Metric) Inner Neck | 137 mm |
Capacity (Metric) | 6 L |
Material | HDPE |
Height (Metric) With Closure | 263 mm |
No. per Pack | 1 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'