missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LARGE Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24307-1-AP-20UL
This item is not returnable.
View return policy
Description
The LARGE gene, which is one of the largest in the human genome, encodes a member of the N-acetylglucosaminyltransferase gene family. It encodes a glycosyltransferase which participates in glycosylation of alpha-dystroglycan, and may carry out the synthesis of glycoprotein and glycosphingolipid sugar chains. It may also be involved in the addition of a repeated disaccharide unit. Mutations in this gene cause MDC1D, a novel form of congenital muscular dystrophy with severe mental retardation and abnormal glycosylation of alpha-dystroglycan. Alternative splicing of this gene results in two transcript variants that encode the same protein.Specifications
| LARGE | |
| Polyclonal | |
| Unconjugated | |
| LARGE1 | |
| KIAA0609, LARGE, LARGE1, like glycosyltransferase, MDC1D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16795, 9215 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95461, Q9Z1M7 | |
| Large1 | |
| LARGE Fusion Protein Ag19458 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction