missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LARGE Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578393
This item is not returnable.
View return policy
Description
Positive Control: mouse liver Predicted Reactivity: Human (100%), Rhesus Monkey (100%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The LARGE gene, which is one of the largest in the human genome, encodes a member of the N-acetylglucosaminyltransferase gene family. It encodes a glycosyltransferase which participates in glycosylation of alpha-dystroglycan, and may carry out the synthesis of glycoprotein and glycosphingolipid sugar chains. It may also be involved in the addition of a repeated disaccharide unit. Mutations in this gene cause MDC1D, a novel form of congenital muscular dystrophy with severe mental retardation and abnormal glycosylation of alpha-dystroglycan. Alternative splicing of this gene results in two transcript variants that encode the same protein.
Specifications
| LARGE | |
| Polyclonal | |
| Unconjugated | |
| Large1 | |
| Acetylglucosaminyltransferase-like 1A; acetylglucosaminyltransferase-like protein; Beta-1,3-glucuronyltransferase LARGE; BPFD#36; enr; eosinophil granule major basic protein 1; fg; froggy; glycosyltransferase-like 1A; Glycosyltransferase-like protein; glycosyltransferase-like protein LARGE1; glycosyltransferase-like protein LARGE1; LARGE xylosyl- and glucuronyltransferase 1; Gyltl1a; KIAA0609; large; LARGE xylosyl- and glucuronyltransferase 1; Large1; like-acetylglucosaminyltransferase; like-glycosyltransferase; MBP1; Mbp-1; MDC1D; MDDGA6; MDDGB6; mKIAA0609; myd; myodystrophy; prepro major basic protein 1; Xylosyltransferase LARGE | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16795, 361368, 9215 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O95461, Q9Z1M7 | |
| Large1 | |
| Recombinant protein encompassing a sequence within the center region of human LARGE. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction