missing translation for 'onlineSavingsMsg'
Learn More

LAR/PTPRF Antibody (384727) [DyLight 350], Novus Biologicals™

Product Code. 30731662 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731662 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731662 Supplier Novus Biologicals Supplier No. FAB3004D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

LAR/PTPRF Monoclonal antibody specifically detects LAR/PTPRF in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAR/PTPRF
Applications Western Blot
Classification Monoclonal
Clone 384727
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias LCA-homolog, protein tyrosine phosphatase, receptor type, F, receptor type, F polypeptide, receptor-linked protein-tyrosine phosphatase LAR, receptor-type tyrosine-protein phosphatase F
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human LAR, Ala27-Glu1251, Accession # NP_569707
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5792
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.