missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAPTM5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523585
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine based on sequence homology.
This gene encodes a transmembrane receptor that is associated with lysosomes. The encoded protein, also known as E3 protein, may play a role in hematopoiesis.
Specifications
| LAPTM5 | |
| Polyclonal | |
| Unconjugated | |
| LAPTM5 | |
| CD40-ligand-activated specific transcripts; CLAST6; gcd-10; granule cell death-10; human retinoic acid-inducible E3 protein; KIAA0085; LAPTM5; lysosomal associated multispanning membrane protein 5; lysosomal multispanning membrane protein 5; lysosomal protein transmembrane 5; lysosomal-associated multitransmembrane protein 5; lysosomal-associated protein transmembrane 5; lysosomal-associated transmembrane protein 5; Retinoic acid-inducible E3 protein | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7805 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q13571 | |
| LAPTM5 | |
| KLH conjugated synthetic peptide between 23-49 amino acids from the N-terminal region of human LAPTM5. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction