missing translation for 'onlineSavingsMsg'
Learn More

Lapatinib, MedChemExpress

Product Code. p-200096742
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
50mg
This item is not returnable. View return policy

Product Code. 16444047

Brand: Medchem Express HY50898

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lapatinib (GW572016) is a potent inhibitor of the ErbB-2 and EGFR tyrosine kinase domains with IC50 values against purified EGFR and ErbB-2 of 10.2 and 9.8 nM, respectively.

Lapatinib (GW572016) is a potent inhibitor of the ErbB-2 and EGFR tyrosine kinase domains with IC50 values against purified EGFR and ErbB-2 of 10.2 and 9.8 nM, respectively.

Specifications

Chemical Name or Material Lapatinib
CAS 231277-92-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H26ClFN4O4S
Quantity 50 mg
Synonym GW572016 GW2016
Solubility Information DMSO : 125 mg/mL (215.12 mM; Need ultrasonic)
SMILES O=S(CCNCC1=CC=C(C2=CC=C3C(C(NC4=CC(Cl)=C(C=C4)OCC5=CC(F)=CC=C5)=NC=N3)=C2)O1)(C)=O
Molecular Weight (g/mol) 581.06
Formula Weight 581.06
Percent Purity 99.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.