missing translation for 'onlineSavingsMsg'
Learn More

Lapatinib, MedChemExpress

Product Code. 16444047
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444047 50 mg 50mg
16421936 500 mg 500mg
16417896 100 mg 100mg
16451949 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16444047 Supplier Medchem Express Supplier No. HY50898
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lapatinib (GW572016) is a potent inhibitor of the ErbB-2 and EGFR tyrosine kinase domains with IC50 values against purified EGFR and ErbB-2 of 10.2 and 9.8 nM, respectively.

Lapatinib (GW572016) is a potent inhibitor of the ErbB-2 and EGFR tyrosine kinase domains with IC50 values against purified EGFR and ErbB-2 of 10.2 and 9.8 nM, respectively.

Specifications

Chemical Name or Material Lapatinib
CAS 231277-92-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H26ClFN4O4S
Quantity 50 mg
Synonym GW572016 GW2016
Solubility Information DMSO : 125 mg/mL (215.12 mM; Need ultrasonic)
SMILES O=S(CCNCC1=CC=C(C2=CC=C3C(C(NC4=CC(Cl)=C(C=C4)OCC5=CC(F)=CC=C5)=NC=N3)=C2)O1)(C)=O
Molecular Weight (g/mol) 581.06
Formula Weight 581.06
Percent Purity 99.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.