missing translation for 'onlineSavingsMsg'
Learn More

Lapatinib Ditosylate Monohydrate, TRC

Product Code. 30472764
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30472764 10 mg 10mg
30472462 1 mg 1mg
30468524 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30472764 Supplier Toronto Research Chemicals Supplier No. TRCL17581310MG
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 388082-78-8
Molecular Formula C29 H26 Cl F N4 O4 S . 2 C7 H8 O3 S . H2 O
Molecular Weight (g/mol) 943.48
Synonym 4-Quinazolinamine, N-[3-chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[[[2-(methylsulfonyl)ethyl]amino]methyl]-2-furanyl]-, 4-methylbenzenesulfonate, hydrate (1:2:1), 4-Quinazolinamine, N-[3-chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[[[2-(methylsulfonyl)ethyl]amino]methyl]-2-furanyl]-, bis(4-methylbenzenesulfonate), monohydrate (9CI), Lapatinib ditosylate monohydrate, GW 572016F, GW 572016F
IUPAC Name N-[3-chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[(2-methylsulfonylethylamino)methyl]furan-2-yl]quinazolin-4-amine;4-methylbenzenesulfonic acid;hydrate
SMILES O.Cc1ccc(cc1)S(=O)(=O)O.Cc2ccc(cc2)S(=O)(=O)O.CS(=O)(=O)CCNCc3oc(cc3)c4ccc5ncnc(Nc6ccc(OCc7cccc(F)c7)c(Cl)c6)c5c4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C29H26ClFN4O4S.2C7H8O3S.H2O/c1-40(36,37)12-11-32-16-23-7-10-27(39-23)20-5-8-26-24(14-20)29(34-18-33-26)35-22-6-9-28(25(30)15-22)38-17-19-3-2-4-21(31)13-19;2*1-6-2-4-7(5-3-6)11(8,9)10;/h2-10,13-15,18,32H,11-12,16-17H2,1H3,(H,33,34,35);2*2-5H,1H3,(H,8,9,10);1H2
Formula Weight 942.1841
Chemical Name or Material Lapatinib Ditosylate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.