missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Lapatinib

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Lapatinib is a potent and selective dual ErbB2 (HER-2) and EGFR inhibitor (IC50 values are 9.8 and 10.2 nM, respectively). Selective for EGFR and ErbB-2 versus other proliferative kinases. Inhibits proliferation in multiple cancer cell lines (average IC50 values <0.16 μM). Exhibits 100-fold average selectivity for the tumor cells versus human foreskin fibroblasts. Inhibits EGFR/ErbB2 autophosphorylation and phosphorylation of the downstream modulator AKT.
Specifications
Specifications
| Chemical Name or Material | N-[3-Chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-[5-[[[2-(methylsulfonyl)ethyl]amino]methyl]-2-furanyl]-4-quinazolinamine |
| CAS | 231277-92-2 |
| Quantity | 50 mg |
| Molecular Formula | C29H26ClFN4O4S |
| Formula Weight | 581.06 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction