missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556207
This item is not returnable.
View return policy
Description
Immunogen sequence: QENWHEGKEN IRAAVAAGCR QIQDLELSSV EVDPCGDAQA AAEGAVLGLY EYDDLKQKKK MAVSAKLYGS GDQEAWQKGV LFASGQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Leucine Aminopeptidase 3 (LAP3) is involved in the processing and regular turnover of intracellular proteins. It catalyzes the removal of unsubstituted N-terminal amino acids from various peptides. Release of an N-terminal amino acid, Xaa-/-Yaa-, in which Xaa is preferably Leu, but may be other amino acids including Pro although not Arg or Lys, and Yaa may be Pro. Amino acid amides and methyl esters are also readily hydrolyzed, but reated on arylamides are exceedingly low.
Specifications
| LAP3 | |
| Polyclonal | |
| Unconjugated | |
| LAP3 | |
| 2410015L10Rik; AA410100; Cytosol aminopeptidase; epididymis secretory protein Li 106; HEL-S-106; Lap; Lap3; LAP-3; LAPEP; leucine aminopeptidase 3; Leucyl aminopeptidase; Pep7; Pep-7; Peps; Pep-S; peptidase 7; peptidase S; Proline aminopeptidase; prolyl aminopeptidase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51056, 66988 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P28838, Q9CPY7 | |
| LAP3 | |
| Recombinant protein corresponding to Human LAP3. Recombinant protein control fragment (Product #RP-95810). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction