missing translation for 'onlineSavingsMsg'
Learn More

LAP1B Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18423724 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423724 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423724 Supplier Novus Biologicals Supplier No. NBP119123AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LAP1B Polyclonal antibody specifically detects LAP1B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAP1B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp586G011, FLJ13142, lamina associated polypeptide 1B, lamina-associated polypeptide 1B, LAP1B, MGC3413, torsin A interacting protein 1, torsin-1A-interacting protein 1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 275 and 325 of human lamina associated polypeptide 1B using the numbering given in entry NP_056417.2 (GeneID 26092).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26092
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.