missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAP1 Antibody (RL13) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB120-2737JF669
This item is not returnable.
View return policy
Description
LAP1 Monoclonal antibody specifically detects LAP1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| LAP1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| lamina-associated polypeptide 1B, torsin A interacting protein 1 | |
| Pore complex-lamina fraction isolated from rat liver nuclear envelopes. | |
| 0.2 mL | |
| Apoptosis, Immunology, Innate Immunity, Zinc Finger | |
| 208263 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| RL13 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction