missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAP (TGF-beta 1) Antibody (27235) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB246MFV500
This item is not returnable.
View return policy
Description
LAP (TGF-beta 1) Monoclonal antibody specifically detects LAP (TGF-beta 1) in Human samples. It is validated for Neutralization,Immunocytochemistry
Specifications
| LAP (TGF-beta 1) | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LAP (TGF-β1) | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction | |
| 7040 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Neutralization, Immunocytochemistry | |
| 27235 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction